Ultrapure Straight Plug Stoppers, Wheaton
Straight Plug stopper, 47 durometer hardness, Gray Bromobutyl, 13×20 mm
Supplier #: W224100-405
Manufactured from an ultrapure bromobutyl formulation with extremely high chemical purity and low gas permeability.
- Low particulate content and low level of extractables
- Free of latex, nitrosamines, and 2-MCBT (2- mercaptobenzothiazole)
- Ideal for "ready for sterilization" and "ready for use" applications
- Autoclavable
Certifications: Complies with US, European, and Japanese Pharmacopeias.
Documents
- Description:Straight Plug stopper, 47 durometer hardness
- Material:Gray Bromobutyl
- I.D.×O.D.:13×20 mm