Liquid-Filled Magnetic Compass
Liquid-Filled Magnetic Compass
Catalog # 470221-936
Supplier: UNITED SCIENTIFIC SUPPLIES
Número de proveedor #: CLF045
Jump to:
Product Details & Documents
Compass for everyday applications.
- Durable
- Liquid-filled
Clear magnetic compass is housed in a black plastic rotatable bezel. Graduated in 5° increments. Outer diameter of bezel is 45mm. Outer diameter of clear compass is 35mm. Compass includes a lanyard.
Especificaciones
- Description:Liquid-Filled Magnetic Compass