ContactUsLinkComponent
Pitchers with Molded Graduations, Globe Scientific
Pitchers with Molded Graduations, Globe Scientific
Catalog # 75838-762
Supplier #: 601160-1
undefined
Pitchers with Molded Graduations, Globe Scientific
Catalog # 75838-762
Supplier Number:  601158-1
Supplier #: 601160-1
Log in for availability
Select an Option
5000 ml
Select an Option
75838-762
Select an Option
601160-1
About this item
These pitchers are ideal for all general laboratory procedures.
  • Molded in handle with thumb grip
  • Special no-drip pour spout
  • Graduated in milliliters (ml)
In one pitcher you can mix, measure and prepare solutions. The pitchers have a no-drip pour spout and a molded-in handle with thumb grip. They are made from heavy-duty plastic and withstands repeated autoclaving. Switching from glass beakers to Globe’s plastic equivalents will greatly reduce breakage and the potential for injury.
Frequently Bought Together