Eisco® Folding Quadrat
Eisco® Folding Quadrat, 50 cm, 50 cm
Catalog # 470220-788
Supplier: EISCO SCIENTIFIC LLC
Número de proveedor #: BI0504
Jump to:
Product Details & Documents
Hands-on field sampling tool.
- Square sampling quadrat
- Folds flat when not in use
- Used for sampling plants and slow moving organisms in the field
- Aligned with GLOBE protocols
For population counts and other measurements in the field. Consists of four white coated steel rods 0.5 m long with markers at 0.25 m, which simply bolt together to form a quadrant frame.
Especificaciones
- Width:50 cm
- Description:Eisco® Folding Quadrat
- Length:50 cm