ContactUsLinkComponent
Helical Spring and Mass Set
Helical Spring and Mass Set
  470159-576
undefined
Helical Spring and Mass Set
  470159-576
 :  PH0325
  • Descripción:
    Helical Spring and Mass Set
  • Referencia:
    470159-576
  • Nº de proveedor:
    PH0325
Demonstrate the extension of a spring in relation to the force applied to it.
  • Includes 1 steel spring and 3 masses