Orders LinkContactUsLinkComponent
Collapsible Metal
Collapsible Metal
Certificates
Collapsible Metal
Collapsible Metal
3 Options Available Below
Product Details & Documents
Dramtically demonstrate the power of atmospheric pressure
  • Demonstrate ideal gas law in a visually spectacular manner
  • Great for classrooms of all sizes
This metal can with stopper and fitting provides an intriguing demonstration of physics principles relating to gas and air pressure. The unit can be purchased including the hand pump and replacement cans are available as well. The can also can be collapsed using boiling water to evacuate air from the can. Includes cap as well as stopper and hose fitting.
Product Family Options
Información del producto
  • Availability
    Price
  • 470222-634
    Collapsible Metal Can with Stopper
    Collapsible Metal Can with Stopper
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $962.60
    $962.60
    Each (1UN)
       $962.60
    Supplier Number: PH0201602
  • 470222-636
    Collapsible Metal Can with Hand Pump and Stopper
    Collapsible Metal Can with Hand Pump and Stopper
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,829.80
    $1,829.80
    Each (1UN)
       $1,829.80
    Supplier Number: PH0201603
  • 470222-638
    Replacement Metal Can with Metal Cap
    Replacement Metal Can with Metal Cap
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $613.50
    $613.50
    Each (1UN)
       $613.50
    Supplier Number: METALCAN