Reversed Phase, J’sphere ODS-H80, HPLC Columns, YMC AMERICA, INC
97031-114
- Formato de columna:Analytical
- Ø int.:4.60 mm
- Longitud:150 mm
- Environmentally Preferable:
- Referencia:97031-114
- Nº de proveedor:JH08S04-1546WT
J’sphere ODS-H80 has a hydrophobic stationary phase.
- Yields a high carbon content and providing a strong, dominant, lipophilic interaction with the nonpolar sites of the sample molecules
- Hydrophobic stationary phase
- Densely covered with polymeric bonded C18 groups
J’sphere ODS-H80 has a hydrophobic stationary phase. It is densely covered with polymeric bonded C18 groups yielding a high carbon content and providing a strong, dominant, lipophilic interaction with the nonpolar sites of the sample molecules. The ability to form H-bonding gives additional selectivity, which is essential for difficult separations, such as drug and corresponding metabolite discrimination.