ContactUsLinkComponent
Hettich UNIVERSAL 320 General Purpose Benchtop Centrifuges, Hettich Lab Technology
Hettich UNIVERSAL 320 General Purpose Benchtop Centrifuges, Hettich Lab Technology
Certificates
undefined
Hettich UNIVERSAL 320 General Purpose Benchtop Centrifuges, Hettich Lab Technology
Specifications
  • Catalog No:
  • 10813-188
  • Detalles eléctricos:
  • 120V
  • Control de temperatura:
  • from -20 to +40°C (320 R)
  • Altura:
  • 13.6” (320), 13.6” (320 R)
  • Profundidad:
  • 20.5” (320), 27.4” (320 R)
  • Ancho:
  • 15.6” (320), 15.8” (320 R)