Orders LinkContactUsLinkComponent
Detonox™ Ultimate Precision Cleaner
Detonox™ Ultimate Precision Cleaner
4×3.8 L (1 gal.), Bottle
Catalog # 75801-040
Supplier:   ALCONOX INC
Número de proveedor #:  2301
Detonox™ Ultimate Precision Cleaner
Catalog # 75801-040
Supplier:  ALCONOX INC
Detonox™ Ultimate Precision Cleaner
Detonox™ Ultimate Precision Cleaner
4×3.8 L (1 gal.), Bottle
Price
$2,932.71
Log in Para disponibilidad
Each (1UN) Case de 4 (1UN)
Product Details & Documents
Detonox is a non-caustic detergent for exceptional removal of botanical, cosmetic, and pharmaceutical polymers, sticky extracts, resins, creams and lotions. Concentrated, highly emulsifying and penetrating formula is free rinsing, and leaves no residues.
  • Concentrated
  • Exceptional cosmetic and pharmaceutical polymer, cream and lotion removal
  • Non-caustic for greater worker safety and greater material compatibility
  • Free rinsing to give you reliable results and no interfering residues
Can be used to remove cosmetic bases and formulations, organics, grease, tars, sticky extracts, resins, waxes, fats, oils, titanium and zinc oxides, ointments and lotions.
Recommended for glass, stainless steel, PTFE, ceramic, cement and solvent resistant plastics and metals. Corrosion testing is advisable.
Cleaning method: Soak, circulate under immersions, ultrasonic cleaners, and manual cleaning.
Documents
Especificaciones
  • Size:
    4×3.8 L (1 gal.)
  • Packaging:
    Bottle