Spatulas, Rounded, Micro, Non-Magnetic, Chemglass
Catalog # 80071-664
Supplier: CHEMGLASS INC.
Small Business Enterprise
Specifications
- Descripción:Stainless steel spatula, #304, non-magnetic, rounded ends
- Longitud:20.3 cm (8")
- Referencia:80071-664
- Nº de proveedor:CG-1981-10
Specifications
About this item
Spatulas are made of non-magnetic stainless steel and feature a mirror finish.
- Spatula will pass through a 14/10 or 14/20 outer joint
- Both ends are rounded and are approximately ⁵/₁₆" (8 mm) wide × 2" (51 mm) in length
- Non-magnetic