DECON-QUAT® 200C, Veltek
DECON-QUAT® 200C, 473 mL (16 oz.), Concentrate, simple mix, Non sterile
Supplier: VELTEK ASSOCIATES, INC. MX
Supplier #: DQ200C07-16Z01
Jump to:
Product Details & Documents
DECON-QUAT® 200C is a quaternary ammonium solution for use on hard non-porous environmental surfaces.
- Effective as a broad spectrum hospital disinfectant, fungicide, deodorizer, hard surface disinfectant, non-food contact sanitizer
- Effective against a broad spectrum of bacteria and viruses
- Available as both sterile and non-sterile
- Completely traceable and has been validated for sterility and shelf life
A phosphate-free formulation designed to provide effective cleaning, deodorizing and disinfection in areas where housekeeping is of prime importance in controlling the hazards of cross-contamination. This product is filled in ISO 5 (Grade A/B, former Class 100), filtered at 0.2 microns, and subsequently terminally sterilized to 10-6 sterility assurance level.
Documents
Specifications
- Description:DECON-QUAT® 200C
- Size:473 mL (16 oz.)
- Sterility:Non sterile
- Form:Concentrate, simple mix