ContactUsLinkComponent
Difco™ MIO Medium, BD Diagnostics
  90003-908
 :  
undefined
Difco™ MIO Medium, BD Diagnostics
  90003-908
 :  273520
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Descripción:
    MIO Medium
  • Tamaño:
    500 g
  • Referencia:
    90003-908
  • Nº de proveedor:
    273520
Motility Indole Ornithine (MIO) Medium is a semisolid medium useful in the identification of members of the Enterobacteriaceae.
  • Casein and peptic digest provide nitrogen and amino acids
  • Yeast extract is a source of vitamin B-complex
  • Dextrose is the fermentable carbohydrate
Motility Indole Ornithine Medium was formulated by Ederer and Clark and Oberhofer and Hajkowski for detection of motility, indole and ornithine decarboxylase production in one tube as an aid in the identification of members of the Enterobacteriaceae family.