Orders LinkContactUsLinkComponent
Difco™ Neutralizing Buffer, BD Life Sciences
Difco™ Neutralizing Buffer, BD Life Sciences
Neutralizing Buffer, 100 g
Catalog # 90002-694
Número de proveedor #:  236210
Difco™ Neutralizing Buffer, BD Life Sciences
Catalog # 90002-694
CAS Number:  
Difco™ Neutralizing Buffer, BD Life Sciences
Difco™ Neutralizing Buffer, BD Life Sciences
Neutralizing Buffer, 100 g
CAS Number:  
Price
$6,190.00
Log in Para disponibilidad
Each (100g)
Product Details & Documents
Neutralizing Buffer is recommended for detection of microorganisms found on dairy and food equipment disinfected with chlorine or quaternary ammonium compounds.
  • Inactivates bactericidal and bacteriostatic effects of chlorine and ammonium
  • Light amber appearance when prepared
  • Recommended for digestion and decontamination of mycobacterial specimens
Monopotassium phosphate provides the buffering capability. Sodium thiosulfate inactivates the effect of chlorine compounds. The aryl sulfonate complex neutralizes the effects of quaternary ammonium compounds.
Documents
Especificaciones
Especificaciones
  • Description:
    Neutralizing Buffer
  • Size:
    100 g