VWR® Basic SPP Coveralls
Acerca de este artículo
Disposable coverall made from spunbond polypropylene (SPP) fabric, offering lightweight and breathable protection for non-hazardous environments. Ideal for general industrial use, food processing, and clean areas requiring basic contamination control.
Lightweight and breathable fabric minimizes heat buildup
- Zipper front closure for easy donning and removal
- Elastic wrists and ankles for secure coverage
- Disposable, single-use design for contamination control
- Economical solution for basic protection needs
- Non-linting material helps maintain cleanliness
- Ideal for general industrial, food processing, and nonhazardous environments
The Basic Protection SPP Coverall is an economical and practical solution for environments requiring lightweight, disposable protective apparel. Constructed from spunbond polypropylene (SPP) fabric, this garment offers a balance of comfort and functionality. The non-woven material is breathable and non-linting, making it suitable for clean areas, food processing, and general industrial applications where contamination control is essential but exposure risks are minimal.
Designed for ease of use, the coverall features a zipper front closure for quick donning and removal, along with elastic wrists and ankles to ensure a secure fit and reduce gaps. The single-use design supports hygiene compliance and simplifies inventory management, while the lightweight construction minimizes heat buildup during extended wear.
Although not intended for hazardous chemical or liquid protection, the Basic Protection SPP Coverall provides an effective barrier against dust, dirt, and dry particulates. This makes it an ideal choice for controlled environments, pre-gowning areas, and routine operations where basic protection is required. By combining cost-efficiency, comfort, and contamination control, this coverall delivers dependable performance for facilities seeking reliable disposable apparel for everyday tasks.
Packaging: Coveralls are packaged 25 per case.
5 Options Available Below
Product Details & Documents
Disposable coverall made from spunbond polypropylene (SPP) fabric, offering lightweight and breathable protection for non-hazardous environments. Ideal for general industrial use, food processing, and clean areas requiring basic contamination control.
Lightweight and breathable fabric minimizes heat buildup
- Zipper front closure for easy donning and removal
- Elastic wrists and ankles for secure coverage
- Disposable, single-use design for contamination control
- Economical solution for basic protection needs
- Non-linting material helps maintain cleanliness
- Ideal for general industrial, food processing, and nonhazardous environments
The Basic Protection SPP Coverall is an economical and practical solution for environments requiring lightweight, disposable protective apparel. Constructed from spunbond polypropylene (SPP) fabric, this garment offers a balance of comfort and functionality. The non-woven material is breathable and non-linting, making it suitable for clean areas, food processing, and general industrial applications where contamination control is essential but exposure risks are minimal.
Designed for ease of use, the coverall features a zipper front closure for quick donning and removal, along with elastic wrists and ankles to ensure a secure fit and reduce gaps. The single-use design supports hygiene compliance and simplifies inventory management, while the lightweight construction minimizes heat buildup during extended wear.
Although not intended for hazardous chemical or liquid protection, the Basic Protection SPP Coverall provides an effective barrier against dust, dirt, and dry particulates. This makes it an ideal choice for controlled environments, pre-gowning areas, and routine operations where basic protection is required. By combining cost-efficiency, comfort, and contamination control, this coverall delivers dependable performance for facilities seeking reliable disposable apparel for everyday tasks.
Packaging: Coveralls are packaged 25 per case.
Documents
Specifications for Product Family
Especificaciones
- Catalog No:
- 414004-337
- 414004-338
- 414004-339
- 414004-340
- 414004-341
- Fabric:
- Spunbound polypropylene
- Spunbound polypropylene
- Spunbound polypropylene
- Spunbound polypropylene
- Spunbound polypropylene
- Closure type:
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Cleanroom class:
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- Pockets:
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Garment application:
- Cleanroom|Industrial|Laboratory
- Cleanroom|Industrial|Laboratory
- Cleanroom|Industrial|Laboratory
- Cleanroom|Industrial|Laboratory
- Cleanroom|Industrial|Laboratory
- Particle shedding (Helmke Drum):
- Level II
- Level II
- Level II
- Level II
- Level II
- Water resistance:
- First line barrier
- First line barrier
- First line barrier
- First line barrier
- First line barrier
- Weight:
- 48 g/m²
- 48 g/m²
- 48 g/m²
- 48 g/m²
- 48 g/m²
Especificaciones
Product Family Options
48 g SPP coveralls
Información del producto
- ColorClothing SizeLengthWaist SizeAvailabilityPrice
- White4XL43" (body), 32" (leg)32"Case (25UN)$1,099.61Paleta de 24 (25UN)$1,198.58Especificaciones:
More
Specifications
Cat. No.414004-337FabricSpunbound polypropyleneClosure typeZip closureCleanroom class10000/ISO 7PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongGarment applicationCleanroom|Industrial|LaboratoryParticle shedding (Helmke Drum)Level IIWater resistanceFirst line barrierWeight48 g/m² - WhiteXXL40½" (body), 31" (leg)29¾"Case (25UN)$1,052.33Paleta de 24 (25UN)$1,147.04Especificaciones:
More
Specifications
Cat. No.414004-338FabricSpunbound polypropyleneClosure typeZip closureCleanroom class10000/ISO 7PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongGarment applicationCleanroom|Industrial|LaboratoryParticle shedding (Helmke Drum)Level IIWater resistanceFirst line barrierWeight48 g/m² - WhiteXL40" (body), 30" (leg)27"Case (25UN)$963.65Paleta de 24 (25UN)$1,050.38Especificaciones:
More
Specifications
Cat. No.414004-339FabricSpunbound polypropyleneClosure typeZip closureCleanroom class10000/ISO 7PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongGarment applicationCleanroom|Industrial|LaboratoryParticle shedding (Helmke Drum)Level IIWater resistanceFirst line barrierWeight48 g/m² - WhiteM37½" (body), 28½" (leg)24¼"Case (25UN)$937.31Paleta de 24 (25UN)$1,021.67Especificaciones:
More
Specifications
Cat. No.414004-340FabricSpunbound polypropyleneClosure typeZip closureCleanroom class10000/ISO 7PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongGarment applicationCleanroom|Industrial|LaboratoryParticle shedding (Helmke Drum)Level IIWater resistanceFirst line barrierWeight48 g/m² - WhiteL38¾" (body), 29½" (leg)25"Case (25UN)$955.60Paleta de 24 (25UN)$1,041.61Especificaciones:
More
Specifications
Cat. No.414004-341FabricSpunbound polypropyleneClosure typeZip closureCleanroom class10000/ISO 7PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongGarment applicationCleanroom|Industrial|LaboratoryParticle shedding (Helmke Drum)Level IIWater resistanceFirst line barrierWeight48 g/m²