Mobile Phase Delivery Bottles, Non-Coated, Microsolv
Supplier: MICROSOLV TECH CORP
Certificates
About this item
These mobile phase delivery bottles are a low cost option for solvent bottles made from Type-33 borosilicate glass.
- Chemically stable
- Non-coated and reusable
- Also useful as media bottles
Excellent for HPLC and GC mobile phase reservoirs. Due to their superior chemical stability, these bottles are useful for storage and delivery of your reagents and solvents that are both aqueous and/or organic in nature.
Delivery: Supplied with white enamel markings, a solid blue cap and a drip free sealing ring. Other color caps are available.
Frequently Bought Together