Orders LinkContactUsLinkComponent
Support Rings, Wire, Humboldt
Support Rings, Wire, Humboldt
Supplier:   HUMBOLDT MFG CO
Support Rings, Wire, Humboldt
Supplier:  HUMBOLDT MFG CO
Certificates
Support Rings, Wire, Humboldt
2 Options Available Below
Product Details & Documents
Formed, bright basic wire circle is welded to shank (except 2" size)
Aluminum muff. Attaches to support rods up to 3/4" (19 mm) in diameter.
Product Family Options
Product Information
  • Ø int.
    Ext.Ø
    Availability
    Price
  • 89370-312
    Size: 2"
    Size: 2"
    4.8 cm
    6.0 cm
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $294.30
    $294.30
    Each (1UN)
       $294.30
    Supplier Number: H-18325
  • 89370-314
    Size: 3"
    Size: 3"
    6.7 cm
    7.9 cm
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $300.09
    $300.09
    Each (1UN)
       $300.09
    Supplier Number: H-18335