Support Rings with Clamps, Chemglass
Catalog # 10034-446
Supplier: CHEMGLASS INC.
Small Business Enterprise
Specifications
- Descripción:Support Rings With Clamps, Small
- Referencia:10034-446
- Diámetro interno:4.4 cm (1.7")
- Nº de proveedor:CG-3057-10
Specifications
About this item
Solid ring with built in clamp for attachment directly to support rod.
- Nickel plated zinc alloy
- Overall length: 180 mm