Twin Bucket, Contec®
Twin divided bucket with lid, stainless steel wringer and casters, Green, 20 L (5.2 gal.)
Supplier: CONTEC, INC. MX
Número de proveedor #: 2743S
Jump to:
Product Details & Documents
The Twin Bucket is a divided bucket with a curved stainless steel wringer.
- Ideal for smaller controlled environments and labs with limited storage space
- Designed to work with the curved stainless steel wringer
- Fully autoclavable (casters are non-autoclavable)
The Twin Bucket is made of durable polypropylene, with a well -fitting lid to eliminate liquid spillage during transport. Use one side of the bucket for cleaning and disinfecting solutions, and the other for rinsing.
Delivery information: Includes 4 casters for added mobility.
Especificaciones
- Capacity:20 L (5.2 gal.)
- Color:Green
- Description:Twin divided bucket with lid, stainless steel wringer and casters