Qualatex® XC 7C Anti-Static Latex Fingercots, QRP
Catalog # 56614-018
Supplier: PIP
Specifications
- Color:Pink
- Descripción:Powder-free
- Longitud:7 cm (2¹³⁄₁₆")
- Tamaño:M
- Grosor:3.5 mil.
- Referencia:56614-018
- Nº de proveedor:7CM
Specifications
About this item
The QRP 7C Anti-Static Clean Room Latex Finger Cots are recommended for use with static sensitive devices.
- Packaged for ISO 5 (Class 100) applications
- Compliant with ASTM and IES-RP-CC 005.3 standards
- Average surface resistance of 5 X 10¹² ohms per square unit
- For use with Class II static sensitive devices (thresholds above 1000V)
- Unique, Stand-Up pouch - easy product identification
7C pink Anti-Static Clean Room Latex Finger Cots are recommended for use with static sensitive devices. The QRP 7C gloves are packaged for ISO 5 (Class 100) applications and are compliant with ASTM and IES-RP-CC 005.3 standards. Finger cots are packaged in gas-impremeable film flushed with semiconductor grade N2. Unique, Stand-Up Pouch provides easy product identification.
Certifications: ESD Safe to ANSI/ESD S20.20-2014.
Caution: This product contains natural rubber latex which may cause allergic reactions.