Boreal Science Microscope Slides
470005-638
- Descripción:Boreal science slides
- Referencia:470005-638
- Nº de proveedor:MSL3X1
Standard Sized Microscope Slides
- Quality slide making supplies
- Traditional size and material
Designed for precise microscopic work, these precleaned slides have edges that are ground smooth, and are made from high-quality, non-corrosive, colorless glass.