BioClean™ Vijon™ Sterile Single Use Goggles, Ansell
75839-022
- Descripción:Vijon™ sterile single use goggles
- Referencia:75839-022
- Nº de proveedor:BVGS
BioClean™ Vijon™ sterile single use goggles are lightweight and non-linting.
- Lightweight PVC frame
- Indirect ventilation system to maintain user comfort
- Optically correct lens
- Toughened polycarbonate, anti-fog, and anti-scratch lens
- Non-linting, latex-free head band offering unrivaled comfort
- ISO Class 4 compatible
BioClean™ Vijon™ sterile single use goggles are constructed from lightweight, ultra-soft PVC. They may also be worn over eyeglasses.
: Manufactured in a facility holding ISO 9001:2008 and 89/686/EEC Article 11b certifications.
Processed in an NEBB certified ISO Class 4 cleanroom environment.
Conforms to Category 2 - Personal Protective Equipment PPE Directive 89/686/EEC.
Complies with requirements of EN 166:2001 standard for personal eye protection.
Tested to ANSI/ISEA Z87.1-2010, the American National Standard for Occupational and Educational Personal Eye and Face Protection Devices.
Processed in an NEBB certified ISO Class 4 cleanroom environment.
Conforms to Category 2 - Personal Protective Equipment PPE Directive 89/686/EEC.
Complies with requirements of EN 166:2001 standard for personal eye protection.
Tested to ANSI/ISEA Z87.1-2010, the American National Standard for Occupational and Educational Personal Eye and Face Protection Devices.
: Goggles are packaged one piece per sealed PE inner bag; 10 inner bags per sealed outer PE bag; six outer bags per lined carton (60 pieces).