BioClean™ Clearview™ Autoclavable Silicone Rubber Goggles, Ansell
Catalog # 75833-999
Supplier: ANSELL HEALTHCARE PRODUCTS LLC MX
Specifications
- Descripción:Clearview™ autoclavable silicone rubber cleanroom goggles
- Referencia:75833-999
Specifications
About this item
BioClean™ Clearview™ Autoclavable Silicone Rubber Goggles are optically correct for clear vision.
- Super-soft silicone rubber frame
- Indirect ventilation system
- Toughened polycarbonate, anti-scratch, optically correct lens
- Adjustable hook and eye clip for easier donning
- Non-linting, latex-free head band
- Tested to withstand >40 autoclave cycles of 30 minutes duration at 121 °C (250 °F)
- Can be worn over eyeglasses
- ISO Class 5 compatible
Constructed in super-soft, lightweight silicone rubber to provide unrivalled comfort, BioClean™ Clearview™ Autoclavable Cleanroom Goggles feature an indirect ventilation system, toughened anti-scratch polycarbonate lens, and can be worn over eyeglasses.
Goggles feature a silicone rubber frame, toughened polycarbonate lens, and latex-free silicone head band with polypropylene hooks.
Provided nonsterile.
Certifications: Manufactured in a facility holding ISO 9001:2008.
Processed in an NEBB certified ISO Class 4 cleanroom environment.
Processed in an NEBB certified ISO Class 4 cleanroom environment.
Packaging: Goggles are packaged one piece per sealed PE bag (to be removed prior to autoclaving); 12 bags per inner box; five boxes per lined carton (60 pieces).