Orders LinkContactUsLinkComponent
Immersion Oil, Type A
Immersion Oil, Type A
Type A Immersion Oil, Low Viscosity, 1 oz.
Catalog # 470201-938
Supplier #:  16482-1OZ
Immersion Oil, Type A
Catalog # 470201-938
CAS Number:  
Immersion Oil, Type A
Immersion Oil, Type A
Type A Immersion Oil, Low Viscosity, 1 oz.
CAS Number:  
Price
$596.56
Log in Para disponibilidad
Each(1oz) Caja de 12 (1 oz)
Product Details & Documents
Essential for Sharp Resolution at High Magnification
  • Critical for clear, sharp magnification
  • Quality slide supplies
A drop of immersion oil between the slide and condenser, and between the slide and objective lens, provides a distortion-free path for light. Safe for all lenses, the low-fluorescence and light-viscosity oil won’t entrain air or dry out the slides or optics. The refractive index is 1.5150, the same as that of glass. From Cargille Labs, the most respected producer of high-quality immersion oil, the oil can easily be cleaned from slides with a bit of xylene on a lens tissue. It comes in an amber bottle with an application rod cap.
Especificaciones
Especificaciones
  • Description:
    Type A Immersion Oil, Low Viscosity
  • Size:
    1 oz.
Especificaciones