Economy Alcohol Burner
Catalog # 470199-936
Supplier: C & A SCIENTIFIC CO INC
Specifications
- Volumen:120 ml (4 oz.)
- Descripción:Alcohol burner
- Referencia:470199-936
- Nº de proveedor:97-5313
Specifications
About this item
Tip-resistant ethyl alcohol burner.
- Nickel-plated snuffer cap extinguishes the flame and seals to prevent evaporation
- Designed to burn 95% Denatured Ethyl Alcohol
Ordering information: Includes one wick. Alcohol is not included.