Fundamentals of Physics Fun Stand
470014-796
- Descripción:Fundamentals of physics fun stand
- Dimensiones:41³/₄×1×1"
- Referencia:470014-796
- Nº de proveedor:FNST08
Ensure accuracy in all your experiments.
- Provide secure support for all of the FUNdamentals of physics kits
- Maximum adaptability in experiments
This solid wood base has a uniquely stable shape, which provides improved accuracy for each experiment.
: Includes instructions, heavy-duty metal pole, 19 holes spaced 5 cm apart (¹/₄" Ø), and Bull's eye level with adjustable rubber pads. For one lab group.