Orders LinkContactUsLinkComponent
Support Ring with Clamp
Support ring with clamp, 8 cm
  470104-526
 :  SRCI03
Support Ring with Clamp
  470104-526
Support Ring with Clamp
Support Ring with Clamp
Support ring with clamp, 8 cm
$215.34
Each (1UN)
Support rings for funnels of all sizes.
  • Cast iron, zinc plated
  • Boss head clamp
Cast iron, zinc-plated support ring with boss head clamp and thumbscrew. Distance from support to center of ring is 4-1/2" for all sizes.
  • Ø:
    8 cm
  • Description:
    Support ring with clamp