Neoprene Burner Tubing
Supplier: EISCO SCIENTIFIC LLC
Certificates
About this item
Connect your burner to a fuel source easily and safely.
- Two sizes available
- Tight secure connection to most common burners and gas sources
This neoprene hose connects your bunsen, tirrill or any other type of burner to a fuel source.
Frequently Bought Together