Meter Stick
Meter stick with plain ends, Hardwood
Catalog # 470046-332
Supplier: AMERICAN SCIENTIFIC LLC SE
Número de proveedor #: 2192-97
Jump to:
Product Details & Documents
7 mm-thick hardwood meter stick has permanently engraved black scales in metric and English.
- 1m (3′) length
One side is graduated in centimeters in 1mm increments. The reverse side is graduated with inches in ⅛″ increments.
Especificaciones
- Description:Meter stick with plain ends
- Material:Hardwood
Accessories
Información del producto
- Catalog #MaterialAvailabilityPrice
- Meter stick with plain ends