Clamp Pulley
Clamp Pulley, 2"
Catalog # 470148-858
Supplier: VWR International
Jump to:
Product Details & Documents
This free-running pulley features a rugged plastic sheave and deep V grooves.
- Pulley Clamps to Surfaces up to One-Inch Thick
- Rugged Plastic Sheave
- Deep V Grooves
- Free Running
It can clamp to surfaces up to one-inch thick.
The sheave is two inches in diameter.
Especificaciones
- Ø:2"
- Description:Clamp Pulley
Accessories
Información del producto
- Catalog #WeightAvailabilityPrice
- Mass Hanger, 500 g Capacity