Orders LinkContactUsLinkComponent
PYREX® Culture Tubes
PYREX® Culture Tubes
   Sustainable Options Available
Supplier:   CORNING CA
PYREX® Culture Tubes
Supplier:  CORNING CA
   Sustainable Options Available
Certificates
PYREX® Culture Tubes
4 Options Available Below
Product Details & Documents
Made of PYREX® borosilicate glass, rimless.
  • Very good chemical resistance
  • Very good temperature resistance
  • Ends are fire-polished
Culture tubes have walls and bottom of a uniform thickness. All tubes feature a white marking spot, with the exception of 6 and 22 mm tubes.
Tube 5 ml can be used for Kahn tests. Tube 9 ml can be used for Wasserman tests.
Specifications for Product Family
Specifications
  • Catalog No:
  • 470211-662
  • 470211-670
  • 470211-672
  • 470211-676
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
Specifications
Product Family Options
Product Information
  • Capacity
    Height
    Ø
    Availability
    Price
  • 9 ml
    100 mm
    13 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Paquete de 72
    Paquete de 72 - $1,520.00
    $1,520.00
    Log in for availability
    Case de 10 (72UN)
    Case de 10 (72UN) - $1,606.88
    $1,606.88
    Paquete de 72
       $1,520.00
    Case de 10 (72UN)
       $1,606.88
    Supplier Number: 9820-13
    Specifications:
    More
    Specifications
    Cat. No.
    470211-662
    Environmentally Preferable
    Environmental certification
  • 27 ml
    150 mm
    18 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Paquete de 72
    Paquete de 72 - $2,420.00
    $2,420.00
    Log in for availability
    Case de 8 (72UN)
    Case de 8 (72UN) - $2,562.95
    $2,562.95
    Paquete de 72
       $2,420.00
    Case de 8 (72UN)
       $2,562.95
    Supplier Number: 9820-18
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    470211-670
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
      Low Manufacturing Impact
  • 34 ml
    150 mm
    20 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Paquete de 72
    Paquete de 72 - $68.80
    $68.80
    Clearance
    Log in for availability
    Case de 8 (72UN)
    Case de 8 (72UN) - $550.40
    $550.40
    Clearance
    Paquete de 72
       $68.80
    Case de 8 (72UN)
       $550.40
    Supplier Number: 9820-20
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    470211-672
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
      Low Manufacturing Impact
  • 70 ml
    200 mm
    25 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Paquete de 48
    Paquete de 48 - $4,040.00
    $4,040.00
    Log in for availability
    Case de 4 (48UN)
    Case de 4 (48UN) - $4,356.74
    $4,356.74
    Paquete de 48
       $4,040.00
    Case de 4 (48UN)
       $4,356.74
    Supplier Number: 9820-25X
    Specifications:
    More
    Specifications
    Cat. No.
    470211-676
    Environmentally Preferable
    Environmental certification