BBL™ Blood Agar Base (Infusion Agar), BD
90000-048
:
- Descripción:BBL™ Blood Agar Base (Infusion Agar)
- Tamaño:500 g
- Referencia:90000-048
- Nº de proveedor:211037
Blood Agar Base (Infusion Agar), with the addition of sterile blood, is used for the isolation, cultivation, and detection of hemolytic activity of streptococci and other fastidious microorganisms.
- All-purpose medium
- Heart muscle infusion, casein peptone, and yeast extract provide nutrients
- Sodium chloride maintains osmotic equilibrium
- Agar is the solidifying agent
Infusion Agar is an all-purpose medium which has been used for many years as a base for the preparation of blood agars. Supplementation with blood (5-10%) provides additional growth factors for fastidious microorganisms, and is the basis for determining hemolytic reactions.