ContactUsLinkComponent
BBL™ Blood Agar Base (Infusion Agar), BD
  90000-048
 :  
BBL™ Blood Agar Base (Infusion Agar), BD
  90000-048
 :  211037
 :  
  • Descripción:
    BBL™ Blood Agar Base (Infusion Agar)
  • Tamaño:
    500 g
  • Referencia:
    90000-048
  • Nº de proveedor:
    211037
Blood Agar Base (Infusion Agar), with the addition of sterile blood, is used for the isolation, cultivation, and detection of hemolytic activity of streptococci and other fastidious microorganisms.
  • All-purpose medium
  • Heart muscle infusion, casein peptone, and yeast extract provide nutrients
  • Sodium chloride maintains osmotic equilibrium
  • Agar is the solidifying agent
Infusion Agar is an all-purpose medium which has been used for many years as a base for the preparation of blood agars. Supplementation with blood (5-10%) provides additional growth factors for fastidious microorganisms, and is the basis for determining hemolytic reactions.