ContactUsLinkComponent
Difco™ Biotin Assay Medium, BD
Difco™ Biotin Assay Medium, BD
Catalog # 90003-264
CAS Number:  
Difco™ Biotin Assay Medium, BD
Catalog # 90003-264
Supplier Number:  241910
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Descripción:
    Difco™ Biotin Assay Medium
  • Tamaño:
    100 g
  • Referencia:
    90003-264
  • Nº de proveedor:
    241910
Specifications
About this item
Biotin Assay Medium is used for determining biotin concentration by the microbiological assay technique.
  • Biotin-free dehydrated medium
  • Addition of biotin gives a growth response
  • Dehydrated appearance: Light beige, homogeneous
Biotin Assay Medium may be used for both turbidimetric and titrimetric analysis. The most effective assay range, using Biotin Assay Medium, has been found to be between 0.1 ng and 1 ng biotin.