Difco™ Baird-Parker Agar Base, BD
Catálogo # 90003-974
Supplier: BECTON DICKINSON DE MEXICO, S. MX
CAS Number:
Specifications
- Descripción:Difco™ Baird-Parker Agar Base
- Tamaño:500 g
- Referencia:90003-974
- Nº de proveedor:276840
Specifications
About this item
Baird-Parker Agar Base is used with EY (Egg Yolk) Tellurite Enrichment in the preparation of Egg-Tellurite-Glycerine-Pyruvate Agar (ETGPA) for selective isolation and enumeration of coagulase-positive staphylococci from food, skin, soil, air, and other materials.
- Peptone, beef extract, and yeast extract provide essential nutrients
- Sodium pyruvate stimulates growth of S. aureus
- Egg yolk aids identification by demonstrating lecithinase activity
- Glycine and lithium chloride inhibit growth of contaminant organisms
Baird-Parker Agar Base may also be used for identification of staphylococci on the basis of their ability to clear egg yolk.