Orders LinkContactUsLinkComponent
Difco™ Antibiotic Medium, BD Life Sciences
Difco™ Antibiotic Medium, BD Life Sciences
Difco™ Antibiotic Medium, BD Life Sciences
Certificates
Difco™ Antibiotic Medium, BD Life Sciences
6 Options Available Below
Product Details & Documents
Antibiotic assay media are used for determining antibiotic potency by the microbiological assay technique.
  • Potency is tested by demonstrating its inhibitory effect
Antibiotic medium 1 and antibiotic medium 4 are used in a cylinder plate method for detecting penicillin in nonfat dry milk.
Antibiotic medium 12 is prepared with the grove and randall formula. It is used for the assay of amphotericin B. Recommended for use in preparing test plates for the cylinder plate assay of nystatin and anisomycin.
Antibiotic medium 19 corresponds to the use described in outline of details for official microbiological assays of antibiotics
Cylinder plate assays are used for commercial preparations of antibiotics, as well as in the quantitative determination of antibioitics in body fluids, animal feeds, and other materials.
Certifications: Antibiotic media are prepared according to the specifications of the USP, European Pharmacopeia, and AOAC International.
Documents
Product Family Options
Información del producto
  • Size
    Availability
    Price
  • 90001-890
    Difco™ antibiotic medium 1
    Difco™ antibiotic medium 1
    500 g
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (500g)
    Each (500g) - $5,360.00
    $5,360.00
    Each (500g)
       $5,360.00
    Supplier Number: 226340
  • 90001-920
    Difco™ antibiotic medium 2
    Difco™ antibiotic medium 2
    500 g
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (500g)
    Each (500g) - $5,540.00
    $5,540.00
    Each (500g)
       $5,540.00
    Supplier Number: 227020
  • 90003-292
    Difco™ antibiotic medium 19
    Difco™ antibiotic medium 19
    500 g
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (500g)
    Each (500g) - $5,470.00
    $5,470.00
    Each (500g)
       $5,470.00
    Supplier Number: 243100
  • 90003-390
    Difco™ antibiotic medium 9
    Difco™ antibiotic medium 9
    500 g
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (500g)
    Each (500g) - $5,460.00
    $5,460.00
    Each (500g)
       $5,460.00
    Supplier Number: 246210
  • 90003-392
    Difco™ antibiotic medium 10
    Difco™ antibiotic medium 10
    500 g
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (500g)
    Each (500g) - $6,260.00
    $6,260.00
    Each (500g)
       $6,260.00
    Supplier Number: 246310
    El artículo requiere control de temperatura para almacenamiento y entrega con cargos adicionales. No es elegible para devolución debido a preocupaciones de seguridad y calidad. Considere los requisitos antes de comprar.
  • 90003-548
    Difco™ antibiotic medium 11
    Difco™ antibiotic medium 11
    500 g
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (500g)
    Each (500g) - $4,720.00
    $4,720.00
    Each (500g)
       $4,720.00
    Supplier Number: 259310