Difco™ Oxford Medium, BD Diagnostics
Difco™ Oxford Medium Base, 500 g
Supplier: BECTON DICKINSON DE MEXICO, S. MX
Número de proveedor #: 222530
Difco™ Oxford Medium, BD Diagnostics
Catalog # 90001-654
Supplier: BECTON DICKINSON DE MEXICO, S. MX
CAS Number:
Jump to:
Product Details & Documents
Oxford Medium Base is used to prepare Oxford Medium or Modified Oxford Medium for isolating and differentiating Listeria monocytogenes.
- Listeria species grow over a pH range of 4.4-9.6
- Dehydrated appearance: Tan, free-flowing, homogeneous
- Motility is most pronounced at 20°C
The most common contaminating bacteria found in food sources potentially containing Listeria are streptococci, especially in the enterococci, micrococci and Bacillus species, Escherichia coli, Pseudomonas aeruginosa, and Proteus vulgaris.
Documents
Certificates
Buscar por número de loteEspecificaciones
Especificaciones
- Description:Difco™ Oxford Medium Base
- Size:500 g