Order Entry
Mexico
Orders LinkContactUsLinkComponent
BBL™ Orange Serum Agar, BD Diagnostics

BBL™ Orange Serum Agar, BD Diagnostics

BBL™ Orange Serum Agar, 500 g
Catalog # 90000-204
Número de proveedor #:  211486
BBL™ Orange Serum Agar, BD Diagnostics
Catalog # 90000-204
CAS Number:  
BBL™ Orange Serum Agar, BD Diagnostics

BBL™ Orange Serum Agar, 500 g

CAS Number:  
Price
$10,920.00
Log in Para disponibilidad
Each (500g)
  • El artículo requiere control de temperatura para almacenamiento y entrega con cargos adicionales. No es elegible para devolución debido a preocupaciones de seguridad y calidad. Considere los requisitos antes de comprar.
  • Return Policy
Product Details & Documents

Orange Serum Agar is used for cultivating aciduric microorganisms, particularly those associated with spoilage of citrus products.

  • Peptone provides a source of carbon and nitrogen
  • Orange serum provides acid environment
  • Yeast extract supplies stimulating vitamins
  • Agar is the solidifying agent

Orange Serum Agar is included in recommended methods for examining fruit beverages.

Documents

Especificaciones

Especificaciones

  • Description:
    BBL™ Orange Serum Agar
  • Size:
    500 g

Especificaciones