SP Wilmad-LabGlass Cylindrical Reaction Vessels with Flat Flange, SP Industries
Catalog # 14207-638
Supplier: SP Industries
Specifications
- Capacidad:1000 ml
- Descripción:Reaction vessel, body dia. 4.25"
- Altura:6½"
- Referencia:14207-638
- Para ser utilizado con:Clamp LG-7316-106
- Flange Size:5-3/8" (OD)
- Nº de proveedor:LG-8080-102
Specifications
About this item
These borosilicate glass cylindrical reaction vessels are without constriction at the top and have a flat bottom.
- Sturdy walls
- Tooled flat flange
They feature sturdy walls and a tooled flat flange with a surface ground to assure a tight seal. There is a gently sloping contour at junction of side wall and bottom, thus preventing any weakening that might occur with a sharp shoulder.
Ordering information: Uses LG-7316-106 clamp.