XPR Microbalances, METTLER TOLEDO®
Proveedor: METTLER TOLEDO S.A. DE C.V. MX
Certificados
Acerca de este artículo
METTLER TOLEDO’s new XPR microbalances incorporate a range of innovative new technologies to deliver the highest accuracy in their class and help make your micro-weighing tasks simpler, safer and more secure.
- Capacities from 2.1 g to 10.1 g
- Readabilities from 0.1µg
- Glass draft shield with motorized drive for precise weighing even in unstable environments.
- Compact design with small footprint.
- Fully automatic adjustment using internal weights.
- Built-in level sensor and leveling aid for fast and easy leveling.
- Capacitive color TFT touch screen
- Two-terminal concept simplifies operation, with the the SmartView terminal sitting above the weighing chamber
- LevelControl: Level warning with graphical levelling guide
- Two programmable sensors for hands-free operation of the draft shield (SmartSens) to speed up frequently recurring tasks.
- User management, change history function, and password protection.
- Storage of SOPs in method library, including sample series and tolerances.
To make the most of your valuable resources, XPR microbalances and ultra-microbalances deliver a unique level of precision with exceptionally low minimum weights.
Certificaciones: UL listed and CSA certified.
Información de pedido: Service packages are offered to assure continual reliability and maintain optimum balance performance. Proper installation and initial calibration are the key to ensuring the balance performs properly from day one. A METTLER TOLEDO technician performs installation qualification and operational qualification (IQ/OQ). Contact your representative for Service package details.
Especificaciones
- Catalog No:
- 75832-782
- Interfaz de comunicación:
- RS-232, USB Host, USB Device
- Voltaje:
- 12 VDC ±3%; 2.25 A, max. ripple: 80 mVpp
Especificaciones
Información relacionada
AT-DS_XPR_Micro_EN
Hoja de datos
F&BFOCUS V4 Issue 2 VWR™ Focus Equipment and Instruments, Issue 3
Documentación del producto
Mettler Toledo Dino Trade In VWR CA 2025.pdf Mettler Toledo Dino Trade-In.pdf
Material promocional
Frequently Bought Together