Mixed Protozoa Slide
Mixed Protozoa, Whole mount, Protists
Catalog # 470111-744
Supplier: UNITED SCIENTIFIC SUPPLIES
Número de proveedor #: 100-45
Jump to:
Product Details & Documents
Compare and Contrast Microorganism Features
- Identifying features clearly distinguishable
- Rigorous quality control standards
Many examples from three major phyla: ciliates, flagellates, and amoebae.
Especificaciones
- Type:Protists
- Description:Mixed Protozoa
- Orientation:Whole mount