ContactUsLinkComponent
BBL™ Thioglycollate Medium, Brewer Modified, BD Life Sciences
BBL™ Thioglycollate Medium, Brewer Modified, BD Life Sciences
Catalog # 90000-294
CAS Number:  
BBL™ Thioglycollate Medium, Brewer Modified, BD Life Sciences
Catalog # 90000-294
Supplier Number:  211716
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Descripción:
    BBL™ Thioglycollate Medium, Brewer Modified
  • Tamaño:
    500 g
  • Referencia:
    90000-294
  • Nº de proveedor:
    211716
Specifications
About this item
Thioglycollate Medium, Brewer Modified is used for testing sterility of biological products and for the isolation of aerobic and anaerobic organisms.
  • Modified for sterility testing of products containing mercury preservatives
  • Peptones, beef extract, and yeast extract provide essential nutrients
  • Suitable for luxuriant growth of fastidious microorganisms
  • Very small amount of agar helps in maintaining anaerobic condition
The modified medium contains more thioglycollate and is recommended for sterility testing procedure.