ContactUsLinkComponent
Difco™ Tetrathionate Broth Base, BD Biosciences
Difco™ Tetrathionate Broth Base, BD Biosciences
Catalog # 90000-008
CAS Number:  
Difco™ Tetrathionate Broth Base, BD Biosciences
Catalog # 90000-008
Supplier Number:  210430
CAS Number:  
Specifications
  • Descripción:
    Difco™ Tetrathionate Broth Base
  • Tamaño:
    500 g
  • Referencia:
    90000-008
  • Nº de proveedor:
    210430
Specifications
About this item
Tetrathionate Broth Base is used as a selective enrichment medium for the isolation of Salmonella from feces, urine, foods, and other materials of sanitary importance.
  • Peptones provide nitrogen, vitamins, amino acids, and carbon
  • Oxgall inhibits Gram-positive microorganisms
  • Calcium carbonate neutralizes and absorbs toxic metabolites
Tetrathionate, which is formed in the medium by the addition of the iodine-iodide solution, inhibits the normal intestinal flora of fecal specimens.