Tryptone Glucose Yeast Extract Agar is recommended for the enumeration of bacteria in water, air, milk, and dairy products.
- Glucose is the energy source
- Dehydrated appearance: Cream to yellow, free-flowing
- Firm, comparable with 1.5% Agar gel
Tryptone Glucose Yeast Extract Agar is used as a standard medium for the bacteriological plate count of milk and dairy products.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.