Difco™ Selenite Broth, BD
Catálogo # 90001-932
Proveedor: BECTON DICKINSON DE MEXICO, S. MX
Número CAS:
Especificaciones
- Descripción:Difco™ Selenite Broth
- Tamaño:500 g
- Referencia:90001-932
- Nº de proveedor:227540
Especificaciones
Acerca de este artículo
Selenite broth is used as a selective medium for the isolation of Salmonella species.
- Selective enrichment for the cultivation of Salmonella species
- Must not be autoclaved
- Casein peptone provides essential nitrogenous and carbon compounds
Selenite Broth is used as an enrichment medium for the isolation of Salmonella from feces, urine, water, foods, and other materials of sanitary importance.