Orders LinkContactUsLinkComponent
Difco™ Sabouraud Dextrose Agar, BD
Difco™ Sabouraud Dextrose Agar, BD
Difco™ Sabouraud Dextrose Agar, BD
Certificates
Difco™ Sabouraud Dextrose Agar, BD
2 Options Available Below
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Sabouraud Dextrose Agar is used in qualitative procedures for the cultivation of dermatophytes and other pathogenic and nonpathogenic fungi from clinical and nonclinical specimens.
  • Low pH is favorable for the growth of fungi
  • Dehydrated appearance: Light beige, free-flowing
  • Peptones are the source of nitrogenous growth factors
The addition of antimicrobics further increases the selectivity of the medium.
Documents
Product Family Options
Product Information
  • Size
    Availability
    Price
  • 90000-038
    Difco™ Sabouraud Dextrose Agar
    Difco™ Sabouraud Dextrose Agar
    500 g
    Log in for availability
    Log in for availability
    Each (500g)
    Each (500g) - $3,040.00
    $3,040.00
    Each (500g)
       $3,040.00
    Supplier Number: 210950
  • 90000-258
    Difco™ Sabouraud Dextrose Agar
    Difco™ Sabouraud Dextrose Agar
    2 kg
    Log in for availability
    Log in for availability
    Each (2kg)
    Each (2kg) - $13,220.00
    $13,220.00
    Each (2kg)
       $13,220.00
    Supplier Number: 211661
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.