Orders LinkContactUsLinkComponent
R2A Agar, BD
R2A Agar, BD
R2A Agar, 500 g
  90000-994
Supplier #:  218263
R2A Agar, BD
  90000-994
 :  
R2A Agar, BD
R2A Agar, 500 g
 :  
$5,310.00
Each (500g)
R2A Agar is used for enumerating heterotrophic organisms in treated potable water.
  • Dehydrated appearance: Light beige, flree-flowing
  • Prepared appearance: Light amber, slightly opalescent
  • Improves recovery of chlorine-tolerant bacteria
A low-nutrient medium, R2A Agar is used in combination with a lower incubation temperature and longer incubation time to stimulate the growth of stressed and chlorine-tolerant bacteria.
Documents
  • Description:
    R2A Agar
  • Size:
    500 g