KIMBLE® Filtering Flask with Standard Taper Joint
KT617500-0003
- Capacidad:250 ml
- Descripción:Filtering Flask with Standard Taper Joint, for a 3/8-inch I.D. Tubing
- Referencia:KT617500-0003
- D.E. x Al:85 x 190 mm
- Nº de proveedor:617500-0003
- Connection Size:24/40
The flasks are manufactured from 33 expansion, low extractable borosilicate glass with heavy wall construction, which provides good mechanical strength under vacuum application.
- Standard taper outer joint 24/40
- Serrated hose connector
: Conforms to USP Type I and ASTM E438, Type I, Class A requirements.