Lens and Mirror Set
Lens and Mirror Set
470220-586
Supplier #: PH0201601
Perfect compliment for a ray box.
- Universal mirror
- Frosted lenses
Optics lens/mirror Kit, includes 1, Acrylic Double Convex frosted lens 89 mm long, 1 Acrylic Double Concave frosted lens 86mm long, 1 Universal Stainless steel mirror - can achieve the following mirrors - Plane, concave, convex - Supplied in Plastic box.
- Description:Lens and Mirror Set