Orders LinkContactUsLinkComponent
RT Multi-Position Hotplate Stirrers
RT Multi-Position Hotplate Stirrers
Supplier:   IKA WORKS, INC. MX
RT Multi-Position Hotplate Stirrers
Supplier:  IKA WORKS, INC. MX
Certificates
RT Multi-Position Hotplate Stirrers
1 Options Available Below
Product Details & Documents
5, 10, or 15-position digital magnetic hotplate stirrers, designed for synchronous heating and stirring
  • Homogeneous temperature distribution across the heating plate
  • Adjustable and reversible operation
  • Eco-mode ensures stirring activities do not warm hotplate surface
  • Error code display and easy operation via touch keypad
  • Adjustable speed in increments of 10 min⁻¹
The magnetic coil technology provides noiseless and consistent stirring on all positions. The surface temperature is infinitely adjustable up to 120 °C, producing a maximum medium temperature of 70 °C (depending on the used vessel). Speed remains constant, even when load changes.
Certifications: RoHS compliant.
Ordering information: Unit comes with detachable power cord.
Specifications for Product Family
Especificaciones
  • Catalog No:
  • 75805-606
  • 75805-608
  • 75805-610
  • Model:
  • RT 5
  • RT 10
  • RT 15
  • Plate material:
  • Aluminium alloy
  • Aluminium alloy
  • Aluminium alloy
  • Plate dimensions:
  • 110×495 mm
  • 180×495 mm
  • 270×495 mm
  • Distance between stirring positions:
  • 90 mm
  • 90 mm
  • 90 mm
  • Deviation for individual stirring position:
  • 0%
  • 0%
  • 0%
  • Speed range:
  • 0 - 1000 rpm
  • 0 - 1000 rpm
  • 0 - 1000 rpm
  • Max. stirring capacity H₂O:
  • 400 ml (per position)
  • 400 ml (per position)
  • 400 ml (per position)
  • W×D×H:
  • 120×610×60 mm
  • 120×610×60 mm
  • 280×610×60 mm
  • Electrical:
  • 115 V
  • 115 V
  • 115 V
  • Heat output:
  • 175 W
  • 375 W
  • 580 W
  • Temperature range:
  • max. 70 °C
  • max. 70 °C
  • max. 70 °C
  • Display:
  • LED
  • LED
  • LED
  • Weight:
  • 4 kg
  • 7.5 kg
  • 10.5 kg
Especificaciones
Product Family Options
Información del producto
  • No. of Positions
    Availability
    Price
  • 75805-606
    RT 5 multi-position hotplate stirrer
    RT 5 multi-position hotplate stirrer
    5
    Discontinued
    Supplier Number: 75805-606
    Especificaciones:
    More
    Specifications
    Cat. No.
    75805-606
    Model
    RT 5
    Plate material
    Aluminium alloy
    Plate dimensions
    110×495 mm
    Distance between stirring positions
    90 mm
    Deviation for individual stirring position
    0%
    Speed range
    0 - 1000 rpm
    Max. stirring capacity H₂O
    400 ml (per position)
    W×D×H
    120×610×60 mm
    Electrical
    115 V
    Heat output
    175 W
    Temperature range
    max. 70 °C
    Display
    LED
    Weight
    4 kg
  • 75805-608
    RT 10 multi-position hotplate stirrer
    RT 10 multi-position hotplate stirrer
    10
    Discontinued
    Supplier Number: 75805-608
    Especificaciones:
    More
    Specifications
    Cat. No.
    75805-608
    Model
    RT 10
    Plate material
    Aluminium alloy
    Plate dimensions
    180×495 mm
    Distance between stirring positions
    90 mm
    Deviation for individual stirring position
    0%
    Speed range
    0 - 1000 rpm
    Max. stirring capacity H₂O
    400 ml (per position)
    W×D×H
    120×610×60 mm
    Electrical
    115 V
    Heat output
    375 W
    Temperature range
    max. 70 °C
    Display
    LED
    Weight
    7.5 kg
  • 75805-610
    RT 15 multi-position hotplate stirrer
    RT 15 multi-position hotplate stirrer
    15
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $137,494.74
    $137,494.74
    Each (1UN)
       $137,494.74
    Supplier Number: 3692601
    Especificaciones:
    More
    Specifications
    Cat. No.
    75805-610
    Model
    RT 15
    Plate material
    Aluminium alloy
    Plate dimensions
    270×495 mm
    Distance between stirring positions
    90 mm
    Deviation for individual stirring position
    0%
    Speed range
    0 - 1000 rpm
    Max. stirring capacity H₂O
    400 ml (per position)
    W×D×H
    280×610×60 mm
    Electrical
    115 V
    Heat output
    580 W
    Temperature range
    max. 70 °C
    Display
    LED
    Weight
    10.5 kg