Orders LinkContactUsLinkComponent
8" Test Sieves, W.S. Tyler™
8" Test Sieves, W.S. Tyler™
Supplier:   W.S. Tyler
8" Test Sieves, W.S. Tyler™
Supplier:  W.S. Tyler
Certificates
8" Test Sieves, W.S. Tyler™
8" Test Sieves, W.S. Tyler™
1 Options Available Below
Product Details & Documents
Overall Height - Full: 25/8"; Half: 15/8"
Product Family Options
Half Height
Información del producto
  • Mesh Size
    USA Series
    Availability
    Price
  • 57332-077
    Stainless Frame-Stainless Cloth
    Stainless Frame-Stainless Cloth
    600 µm
    #30
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $4,350.53
    $4,350.53
    Each (1UN)
       $4,350.53
    Supplier Number: 5367