Mole Display Set
Mole Display Set
Catalog # 470220-776
Supplier: EISCO SCIENTIFIC LLC
Número de proveedor #: PHMOL15
Jump to:
Product Details & Documents
Calculate Atomic Volume and Radius.
- Includes 4 different specimens
- Can be used year after year when teaching about moles
- Can be used to calculate specific heat as well
Solid and regular in shape, the specimens in this set provide a clear visual distinction of different sized atoms that represent a mole. Students can handle each specimen for hands-on learning and use them to determine atomic mass, volume, and radius; molar heat capacity; density; and specific heat. Once calculations are completed, atomic mass of unknown elements in the lab can be computed. Includes elements aluminum, zinc, copper, and iron.
Especificaciones
- Description:Mole Display Set